accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112522-64-2

CH3ONHONHH2N

CAS RN 112522-64-2

IUPAC Name: N-(2-aminophenyl)-4-acetamidobenzamide
Synonyms: tacedinaline
Molecular Weight (g/mol): 269.3
Molecular Formula: C15H15N3O2
InChi Key: VAZAPHZUAVEOMC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1N
Molecular Weight (g/mol) 
  • (5)
  • (2)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (5)
Grade 
  • (5)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products