CAS RN 1126-46-1
CAS RN 1126-46-1
IUPAC Name:
methyl 4-chlorobenzoate
Synonyms:
methyl P-chlorobenzoate
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
LXNFVVDCCWUUKC-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Methyl 4-chlorobenzoate, 99%, Thermo Scientific Chemicals
CAS: 1126-46-1 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.59 MDL Number: MFCD00000621 InChI Key: LXNFVVDCCWUUKC-UHFFFAOYSA-N Synonym: methyl p-chlorobenzoate,4-chlorobenzoic acid methyl ester,benzoic acid, 4-chloro-, methyl ester,unii-m0u80j80va,benzoic acid, p-chloro-, methyl ester,methyl-p-chlorobenzoate,p-chlorobenzoic acid methyl ester,methyl-4-chlorobenzoate,4-chloro-benzoic acid methyl ester,decitabine related compound d PubChem CID: 14307 IUPAC Name: methyl 4-chlorobenzoate SMILES: COC(=O)C1=CC=C(Cl)C=C1