accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1128-61-6

H3CFN

CAS RN 1128-61-6

IUPAC Name: 6-fluoro-2-methylquinoline
Synonyms: 6-fluoro-2-methylquinoline
Molecular Weight (g/mol): 161.18
Molecular Formula: C10H8FN
InChi Key: GPIARMSVZOEZCV-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(F)C=CC2=N1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products