accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1129-35-7

CH3OON

CAS RN 1129-35-7

IUPAC Name: methyl 4-cyanobenzoate
Synonyms: methyl 4-cyanobenzoate
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: KKZMIDYKRKGJHG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products