accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112926-02-0

H3COOOONOOOCH3OCH3OONOOOH3COOOOH3CONHOOOH3CO

CAS RN 112926-02-0

IUPAC Name: (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}(2-{2-[5-(6-{[(acetyloxy)methoxy]carbonyl}-1H-indol-2-yl)-2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]phenoxy]ethoxy}-4-methylphenyl)amino)acetate
Synonyms: (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}(2-{2-[5-(6-{[(acetyloxy)methoxy]carbonyl}-1H-indol-2-yl)-2-[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]phenoxy]ethoxy}-4-methylphenyl)amino)acetate
Molecular Weight (g/mol): 1009.92
Molecular Formula: C47H51N3O22
InChi Key: CAWBRCOBJNWRLK-UHFFFAOYSA-N
SMILES: CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)C1=CC=C(C)C=C1OCCOC1=CC(=CC=C1N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C1=CC2=CC=C(C=C2N1)C(=O)OCOC(C)=O
Product Type 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products