accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1131-52-8

CH3OOOCH3

CAS RN 1131-52-8

IUPAC Name: 3-ethoxy-4-methoxybenzaldehyde
Synonyms: 3-ethoxy-4-methoxybenzaldehyde
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: VAMZHXWLGRQSJS-UHFFFAOYSA-N
SMILES: CCOC1=CC(C=O)=CC=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products