accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1131912-76-9

BOOH3CCH3H3CH3CO

CAS RN 1131912-76-9

IUPAC Name: 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 212.1
Molecular Formula: C11H21BO3
InChi Key: NLSMOSUUBUCSPL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products