CAS RN 1135278-41-9
CAS RN 1135278-41-9
IUPAC Name:
1-(2-ethylphenoxy)-3-[(1,2,3,4-tetrahydronaphthalen-1-yl)amino]propan-2-ol hydrochloride
Synonyms:
1-(2-ethylphenoxy)-3-(1,2,3,4-tetrahydronaphthalen-1-ylamino)propan-2-ol hydrochloride
Molecular Weight (g/mol):
361.91
Molecular Formula:
C21H28ClNO2
InChi Key:
SHUCXUIOEAAJJL-UHFFFAOYNA-N
SMILES:
Cl.CCC1=CC=CC=C1OCC(O)CNC1CCCC2=CC=CC=C12
1
–
1
of
1
results
1
Tocris Bioscience™ SR 59230A hydrochloride

CAS: 1135278-41-9 Molecular Formula: C21H28ClNO2 Molecular Weight (g/mol): 361.91