accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113559-13-0

HHClClCH3NOHNSH3COON

CAS RN 113559-13-0

IUPAC Name: dihydrogen N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dichloride
Synonyms: dihydrogen N-(4-{1-[2-(6-methylpyridin-2-yl)ethyl]piperidine-4-carbonyl}phenyl)methanesulfonamide dichloride
Molecular Weight (g/mol): 474.44
Molecular Formula: C21H29Cl2N3O3S
InChi Key: ZQBNWMFBOSOOLX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC=CC(CCN2CCC(CC2)C(=O)C2=CC=C(NS(C)(=O)=O)C=C2)=N1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products