accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1138011-19-4

NNH2OOHHClHCl

CAS RN 1138011-19-4

IUPAC Name: 4-aminopyridine-3-carboxylic acid dihydrochloride
Synonyms: 4-aminopyridine-3-carboxylic acid dihydrochloride
Molecular Weight (g/mol): 211.04
Molecular Formula: C6H8Cl2N2O2
InChi Key: ZCKYJHVCJAUVAE-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1=C(C=NC=C1)C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products
1x1 image pixel for data collection