accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113906-27-7

CH3ONNHOO(E/Z)

CAS RN 113906-27-7

IUPAC Name: N'-[(5-methylfuran-2-yl)methylidene]-2-phenoxybenzohydrazide
Synonyms: N'-[(5-methylfuran-2-yl)methylidene]-2-phenoxybenzohydrazide
Molecular Weight (g/mol): 320.35
Molecular Formula: C19H16N2O3
InChi Key: JJEDWBQZCRESJL-UHFFFAOYSA-N
SMILES: CC1=CC=C(O1)C=NNC(=O)C1=CC=CC=C1OC1=CC=CC=C1
Grade 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products