accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114152-23-7

OHOFFF

CAS RN 114152-23-7

IUPAC Name: 2-(2,3,6-trifluorophenyl)acetic acid
Synonyms: (2,3,6-trifluorophenyl)acetic acid
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: QRAZASHLGLHKEB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=CC(F)=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products