accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1142-20-7

CH3(S)NHOOOHO

CAS RN 1142-20-7

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: TYRGLVWXHJRKMT-QMMMGPOBSA-N
SMILES: C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (2)
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products