CAS RN 114528-79-9
CAS RN 114528-79-9
IUPAC Name:
2-(3,4-dichlorophenyl)-N-methyl-N-[2-(pyrrolidin-1-yl)cyclohexyl]acetamide hydrate hydrochloride
Synonyms:
2-(3,4-dichlorophenyl)-N-methyl-N-[2-(pyrrolidin-1-yl)cyclohexyl]acetamide hydrate hydrochloride
Molecular Weight (g/mol):
423.8
Molecular Formula:
C19H29Cl3N2O2
InChi Key:
UWPRPWMHQDPOFR-UHFFFAOYNA-N
SMILES:
O.Cl.CN(C1CCCCC1N1CCCC1)C(=O)CC1=CC(Cl)=C(Cl)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ (-)-U-50488 hydrochloride

CAS: 114528-79-9 Molecular Formula: C19H29Cl3N2O2 Molecular Weight (g/mol): 423.80