accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1146-99-2

ClOO

CAS RN 1146-99-2

IUPAC Name: 2-(4-chlorophenyl)-2,3-dihydro-1H-indene-1,3-dione
Synonyms: clorindione C15H9ClO2 氯茚二酮
Molecular Weight (g/mol): 256.69
Molecular Formula: C15H9ClO2
InChi Key: NJDUWAXIURWWLN-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products