accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114636-31-6

CH3ONHHN

CAS RN 114636-31-6

IUPAC Name: N-(pyrrolidin-3-yl)acetamide
Synonyms: N-(pyrrolidin-3-yl)acetamide
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: HDCCJUCOIKLZNM-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCNC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products