accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114715-39-8

NH2(R)N

CAS RN 114715-39-8

IUPAC Name: (3R)-1-benzylpyrrolidin-3-amine
Synonyms: (3R)-1-benzylpyrrolidin-3-amine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: HBVNLKQGRZPGRP-LLVKDONJSA-N
SMILES: N[C@@H]1CCN(CC2=CC=CC=C2)C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products