CAS RN 114870-03-0
CAS RN 114870-03-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Fondaparinux sodium, MedChemExpress
MedChemExpress Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor.
