accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115-29-7

ClClClOSOOClClCl

CAS RN 115-29-7

IUPAC Name: 1,9,10,11,12,12-hexachloro-4,6-dioxa-5λ⁴-thiatricyclo[7.2.1.0²,⁸]dodec-10-en-5-one
Synonyms: endosulfan devisulphan pff thiodan 4E caswell no. 420 thifor thiodan 4E insecticide liquid rcra waste number P050 thiotox (insecticide) endosulfan (substance) thionate
Molecular Weight (g/mol): 406.9
Molecular Formula: C9H6Cl6O3S
InChi Key: RDYMFSUJUZBWLH-UHFFFAOYNA-N
SMILES: ClC1=C(Cl)C2(Cl)C3COS(=O)OCC3C1(Cl)C2(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (6)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (4)
  • (2)

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products