CAS RN 115055-85-1
CAS RN 115055-85-1
IUPAC Name:
zinc(2+) (4-bromophenyl)methanide bromide
Synonyms:
zinc(2+) (4-bromophenyl)methanide bromide
Molecular Weight (g/mol):
315.31
Molecular Formula:
C7H6Br2Zn
InChi Key:
HJQDBIKHGQSYJU-UHFFFAOYSA-M
SMILES:
[Zn++].[Br-].[CH2-]C1=CC=C(Br)C=C1