CAS RN 115550-35-1
CAS RN 115550-35-1
IUPAC Name:
7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1,2-diazatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Synonyms:
marbofloxacin 马波沙星
Molecular Weight (g/mol):
362.36
Molecular Formula:
C17H19FN4O4
InChi Key:
BPFYOAJNDMUVBL-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN3N(C)COC1=C23)C(O)=O
1
–
3
of
3
results
1
Marbofloxacin, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Medchem Express Marbofloxacin, MedChemExpress
MedChemExpress Marbofloxacin is a third generation fluoroquinolone and orally active antimicrobial agent, which has a broad spectrum bactericidal activity and good efficacy. Marbofloxacin can be used for the research of infections by Gram-positive and Gram-negative bacteria and Mycoplasma.

3
Honeywell Fluka Marbofloxacin, Fluka™
Analytical standard, reference material