accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115668-57-0

H3COOCH3OOCH3OOY

CAS RN 115668-57-0

IUPAC Name: yttrium(3+) tris(2-methoxyethan-1-olate)
Synonyms: yttrium(3+) tris(2-methoxyethanolate)
Molecular Weight (g/mol): 314.17
Molecular Formula: C9H21O6Y
InChi Key: IVXUPDRNSUCVNX-UHFFFAOYSA-N
SMILES: [Y+3].COCC[O-].COCC[O-].COCC[O-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products