accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116-29-0

ClSOOClClCl

CAS RN 116-29-0

IUPAC Name: 1,2,4-trichloro-5-(4-chlorobenzenesulfonyl)benzene
Synonyms: duphar tetradiphon caswell no. 836 tedane combi tedane tedion tedion V-18 tetradichlone tetrachlorodiphenyl sulfone roztoczol
Molecular Weight (g/mol): 356.04
Molecular Formula: C12H6Cl4O2S
InChi Key: MLGCXEBRWGEOQX-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)C1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
  • (1)
Concentration 
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products