accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1160514-60-2

H3COONNNHOClClClFFF

CAS RN 1160514-60-2

IUPAC Name: ethyl 1-[4-(2,3,3-trichloroprop-2-enamido)phenyl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Synonyms: ethyl 1-[4-(2,3,3-trichloroprop-2-enamido)phenyl]-5-(trifluoromethyl)pyrazole-4-carboxylate
Molecular Weight (g/mol): 456.63
Molecular Formula: C16H11Cl3F3N3O3
InChi Key: RZHGONNSASQOAY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N(N=C1)C1=CC=C(NC(=O)C(Cl)=C(Cl)Cl)C=C1)C(F)(F)F
Color 
  • (4)
Grade 
  • (4)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products