accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1163-36-6

HClClNNN

CAS RN 1163-36-6

IUPAC Name: hydrogen 1-[(4-chlorophenyl)methyl]-2-[(pyrrolidin-1-yl)methyl]-1H-1,3-benzodiazole chloride
Synonyms: hydrogen clemizole chloride
Molecular Weight (g/mol): 362.3
Molecular Formula: C19H21Cl2N3
InChi Key: DNFMJYXRIMLMBZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=C(CN2C(CN3CCCC3)=NC3=CC=CC=C23)C=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products