accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116450-56-7

NOOON

CAS RN 116450-56-7

IUPAC Name: 16-(2-carboxylatophenyl)-3-oxa-9λ⁵,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,4,9(28),13,15,17,19(27)-heptaen-9-ylium
Synonyms: rhodamine 101 rhodamine 640
Molecular Weight (g/mol): 490.6
Molecular Formula: C32H30N2O3
InChi Key: MUSLHCJRTRQOSP-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1=CC=CC=C1C1=C2C=C3CCC[N+]4=C3C(CCC4)=C2OC2=C3CCCN4CCCC(C=C12)=C34
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products