accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116895-09-1

CH3NCH3NNH2NCl

CAS RN 116895-09-1

IUPAC Name: 6-chloro-N4,N4-diethylpyrimidine-2,4-diamine
Synonyms: 6-chloro-N4,N4-diethylpyrimidine-2,4-diamine
Molecular Weight (g/mol): 200.67
Molecular Formula: C8H13ClN4
InChi Key: JZPXSIGRVGTURL-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC(Cl)=NC(N)=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products