man

Test

CAS RN 117-18-0

ONOClClClCl

CAS RN 117-18-0

IUPAC Name: 1,2,4,5-tetrachloro-3-nitrobenzene
Synonyms: 1,2,4,5-tetrachloro-3-nitrobenzene
Molecular Weight (g/mol): 260.88
Molecular Formula: C6HCl4NO2
InChi Key: XQTLDIFVVHJORV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C(Cl)=CC(Cl)=C1Cl
For Use With (Equipment) 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)