accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117-34-0

HOO

CAS RN 117-34-0

IUPAC Name: 2,2-diphenylacetic acid
Synonyms: diphenylacetic acid
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: PYHXGXCGESYPCW-UHFFFAOYSA-N
SMILES: OC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products