accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117048-59-6

CAS RN 117048-59-6

IUPAC Name: 2-methoxy-5-[(1E)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
Synonyms: combretastatin A-4
Molecular Weight (g/mol): 316.35
Molecular Formula: C18H20O5
InChi Key: HVXBOLULGPECHP-AATRIKPKSA-N
SMILES: COC1=CC=C(\C=C\C2=CC(OC)=C(OC)C(OC)=C2)C=C1O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products