CAS RN 1173022-16-6
CAS RN 1173022-16-6
IUPAC Name:
1'-{2-[4-(trifluoromethyl)phenyl]ethyl}-1,2-dihydrospiro[3,1-benzoxazine-4,4'-piperidin]-2-one hydrochloride
Synonyms:
1'-{2-[4-(trifluoromethyl)phenyl]ethyl}-1H-spiro[3,1-benzoxazine-4,4'-piperidin]-2-one hydrochloride
Molecular Weight (g/mol):
426.86
Molecular Formula:
C21H22ClF3N2O2
InChi Key:
KRRISOFSWVKYBF-UHFFFAOYSA-N
SMILES:
Cl.FC(F)(F)C1=CC=C(CCN2CCC3(CC2)OC(=O)NC2=CC=CC=C32)C=C1
1
–
2
of
2
results
1
Tocris RS 102895 hydrochloride, Tocris Bioscience™

CAS: 1173022-16-6 Molecular Formula: C21H22ClF3N2O2 Molecular Weight (g/mol): 426.864 InChI Key: KRRISOFSWVKYBF-UHFFFAOYSA-N PubChem CID: 16759153 IUPAC Name: 1'-[2-[4-(trifluoromethyl)phenyl]ethyl]spiro[1H-3,1-benzoxazine-4,4'-piperidine]-2-one;hydrochloride SMILES: C1CN(CCC12C3=CC=CC=C3NC(=O)O2)CCC4=CC=C(C=C4)C(F)(F)F.Cl
2
Medchem Express RS102895 hydrochloride, MedChemExpress
MedChemExpress RS102895 hydrochloride is a potent CCR2 antagonist, with an IC50 of 360 nM, and shows no effect on CCR1.
