CAS RN 1173022-21-3
CAS RN 1173022-21-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscienceâ„¢ STO-609 acetate
Selective CaM kinase kinase inhibitor
2
STEMCELL Technologies STO-609 (Acetate)
Calcium signaling inhibitor; Inhibits calcium/calmodulin-dependent kinase kinases (CaMKKs)
