accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1173900-33-8

H3CNONCH3HNOHONO

CAS RN 1173900-33-8

IUPAC Name: 2-({1-[7-methyl-2-(morpholin-4-yl)-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl]ethyl}amino)benzoic acid
Synonyms: 2-({1-[7-methyl-2-(morpholin-4-yl)-4-oxopyrido[1,2-a]pyrimidin-9-yl]ethyl}amino)benzoic acid
Molecular Weight (g/mol): 408.46
Molecular Formula: C22H24N4O4
InChi Key: IRTDIKMSKMREGO-UHFFFAOYNA-N
SMILES: CC(NC1=CC=CC=C1C(O)=O)C1=CC(C)=CN2C(=O)C=C(N=C12)N1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products