accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1174064-66-4

ONNBr

CAS RN 1174064-66-4

IUPAC Name: 2-(4-bromo-1H-pyrazol-1-yl)benzaldehyde
Synonyms: 2-(4-bromopyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 251.08
Molecular Formula: C10H7BrN2O
InChi Key: SHWJJFKWHUIAFC-UHFFFAOYSA-N
SMILES: BrC1=CN(N=C1)C1=CC=CC=C1C=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products