CAS RN 117423-74-2
CAS RN 117423-74-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
LY223982, MedChemExpress
MedChemExpress LY223982 is a potent and specific inhibitor of leukotriene B4 receptor, with an IC50 of 13.2 nM against [3H]LTB4 binding to LTB4 receptor.
