accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117464-70-7

CH3OOOOOOOOCH3OOOOOOCH3OOOH3COOOOCH3

CAS RN 117464-70-7

IUPAC Name: (acetyloxy)methyl 3-{3',6'-bis[(acetyloxy)methoxy]-7'-{3-[(acetyloxy)methoxy]-3-oxopropyl}-5-{[(acetyloxy)methoxy]carbonyl}-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl}propanoate
Synonyms: (acetyloxy)methyl 3-{3',6'-bis[(acetyloxy)methoxy]-7'-{3-[(acetyloxy)methoxy]-3-oxopropyl}-5-{[(acetyloxy)methoxy]carbonyl}-3-oxospiro[2-benzofuran-1,9'-xanthen]-2'-yl}propanoate
Molecular Weight (g/mol): 880.76
Molecular Formula: C42H40O21
InChi Key: NTECHUXHORNEGZ-UHFFFAOYSA-N
SMILES: CC(=O)OCOC(=O)CCC1=C(OCOC(C)=O)C=C2OC3=CC(OCOC(C)=O)=C(CCC(=O)OCOC(C)=O)C=C3C3(OC(=O)C4=C3C=CC(=C4)C(=O)OCOC(C)=O)C2=C1
Product Type 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products