accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117467-28-4

H3CSNNOHNONOH3CSNNH2SOOOOCH3H3CCH3(E/Z)(E/Z)

CAS RN 117467-28-4

IUPAC Name: {7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[2-(4-methyl-1,3-thiazol-5-yl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy}methyl 2,2-dimethylpropanoate
Synonyms: {7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[2-(4-methyl-1,3-thiazol-5-yl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy}methyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 620.71
Molecular Formula: C25H28N6O7S3
InChi Key: AFZFFLVORLEPPO-UHFFFAOYNA-N
SMILES: CON=C(C(=O)NC1C2SCC(C=CC3=C(C)N=CS3)=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C1=CSC(N)=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products