accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117620-77-6

H3CONOO

CAS RN 117620-77-6

IUPAC Name: 7-ethoxy-2-oxo-2H-chromene-3-carbonitrile
Synonyms: 7-ethoxy-2-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: YAFGHMIAFYQSCF-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C=C(C#N)C(=O)OC2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products