accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117704-25-3

H3CO(S)(S)O(S)CH3(S)HOO(S)H3CO(R)(S)OH3CO(S)H3CO(R)(R)HO(R)OO(S)(R)CH3O(S)O(R)(S)CH3(S)OHCH3(E)(E)(E)

CAS RN 117704-25-3

IUPAC Name: (1'R,2S,4'S,5S,6R,8'R,10'E,13'S,14'E,16'E,20'R,21'R,24'S)-6-cyclohexyl-21',24'-dihydroxy-12'-{[(2R,4S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one
Synonyms: doramectin
Molecular Weight (g/mol): 899.13
Molecular Formula: C50H74O14
InChi Key: QLFZZSKTJWDQOS-NJPOTTGESA-N
SMILES: CO[C@H]1C[C@H](OC2[C@H](C)O[C@H](C[C@@H]2OC)OC2[C@@H](C)\C=C\C=C3/CO[C@@H]4[C@H](O)C(C)=C[C@@H](C(=O)O[C@H]5C[C@@H](C\C=C2/C)O[C@@]2(C5)O[C@H](C5CCCCC5)[C@@H](C)C=C2)[C@]34O)O[C@@H](C)[C@@H]1O
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products