CAS RN 1177865-17-6
CAS RN 1177865-17-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express NSC 23766 trihydrochloride, MedChemExpress
MedChemExpress NSC 23766 trihydrochloride is an inhibitor of Rac1 activation.
