accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1184-84-5

NaOSOOH2C

CAS RN 1184-84-5

IUPAC Name: sodium ethenesulfonate
Synonyms: sodium apolate 阿朴酸钠
Molecular Weight (g/mol): 130.09
Molecular Formula: C2H3NaO3S
InChi Key: BWYYYTVSBPRQCN-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C=C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products