accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1184172-35-7

H3CONFF

CAS RN 1184172-35-7

IUPAC Name: 2,6-difluoro-4-methoxypyridine
Synonyms: 2,6-difluoro-4-methoxypyridine
Molecular Weight (g/mol): 145.11
Molecular Formula: C6H5F2NO
InChi Key: HRTFKBJLWDISRQ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=NC(F)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products