accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1184918-22-6

FBrBr

CAS RN 1184918-22-6

IUPAC Name: 2-bromo-1-(bromomethyl)-3-fluorobenzene
Synonyms: 2-bromo-1-(bromomethyl)-3-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: ZJSRIMJDFLFPJI-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CBr)=C1Br

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products