accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 118745-63-4

CH3BrF

CAS RN 118745-63-4

IUPAC Name: 1-(bromomethyl)-2-fluoro-4-methylbenzene
Synonyms: 1-(bromomethyl)-2-fluoro-4-methylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: SSDOQXRIONFILC-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(CBr)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products