accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 120-75-2

H3CSN

CAS RN 120-75-2

IUPAC Name: 2-methyl-1,3-benzothiazole
Synonyms: 2-methylbenzothiazole
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: DXYYSGDWQCSKKO-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products