accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 120-87-6

H3C(R)OH(R)OHOHO

CAS RN 120-87-6

IUPAC Name: (9R,10R)-9,10-dihydroxyoctadecanoic acid
Synonyms: 9,10-dihydroxystearic acid
Molecular Weight (g/mol): 316.48
Molecular Formula: C18H36O4
InChi Key: VACHUYIREGFMSP-IAGOWNOFSA-N
SMILES: CCCCCCCC[C@@H](O)[C@H](O)CCCCCCCC(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products