CAS RN 120068-37-3
CAS RN 120068-37-3
IUPAC Name:
5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile
Synonyms:
fipronil regent C12H4cl2F6N4OS
Molecular Weight (g/mol):
437.14
Molecular Formula:
C12H4Cl2F6N4OS
InChi Key:
ZOCSXAVNDGMNBV-UHFFFAOYNA-N
SMILES:
NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
1
–
6
of
6
results
1
Thermo Scientific Chemicals Fipronil
CAS: 120068-37-3 Molecular Formula: C12H4Cl2F6N4OS Molecular Weight (g/mol): 437.14 InChI Key: ZOCSXAVNDGMNBV-UHFFFAOYNA-N IUPAC Name: 5-amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-trifluoromethanesulfinyl-1H-pyrazole-3-carbonitrile SMILES: NC1=C(C(=NN1C1=C(Cl)C=C(C=C1Cl)C(F)(F)F)C#N)S(=O)C(F)(F)F
6
Honeywell Fluka Fipronil, Fluka™
Analytical standard, reference material