accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1207-72-3

CH3NS

CAS RN 1207-72-3

IUPAC Name: 10-methyl-10H-phenothiazine
Synonyms: 10H-phenothiazine, 10-methyl-
Molecular Weight (g/mol): 213.3
Molecular Formula: C13H11NS
InChi Key: QXBUYALKJGBACG-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2SC2=CC=CC=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products