CAS RN 121058-88-6
CAS RN 121058-88-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific™ TheraPure™ Ac-rC Phosphoramidite, 20-400 finish bottle
Thermo Scientific™ TheraPure™ Ac-rC Phosphoramidite, 20-400 finish bottle
Ac-rC CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with Acetyl (Ac) base protection, for use in oligonucleotide synthesis.