accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 122918-25-6

BrNN

CAS RN 122918-25-6

IUPAC Name: 6-bromopyridine-2-carbonitrile
Synonyms: 6-bromopyridine-2-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: HNEBPTAKURBYRM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)C#N
Physical Form 
  • (1)
Quantity 
  • (1)
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products